missing translation for 'onlineSavingsMsg'
Learn More

IU1, Tocris Bioscience™

USP14 inhibitor

Brand:  Tocris (Bio-Techne) 408810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13434124

  • £173.46 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of Usp14; inhibits the catalytic activity of proteasome-associated Usp14 in vitro (IC50 < 4 μM). Stimulates proteasome activity and substrate degradation.
Specifications

Chemical Identifiers

314245-33-5
300.377
iu1, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethan-1-one, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethanone, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-ylethanone, 1-1-4-fluoro-phenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl-ethanone, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-1-pyrrolidinyl ethanone, usp14 inhibitor, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-yl ethanone, iu-1
1-[1-(4-fluorophenyl)-2,5-dimethylpyrrol-3-yl]-2-pyrrolidin-1-ylethanone
C18H21FN2O
JUWDSDKJBMFLHE-UHFFFAOYSA-N
675434
CC1=CC(=C(N1C2=CC=C(C=C2)F)C)C(=O)CN3CCCC3

Specifications

10mg
314245-33-5
iu1, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethan-1-one, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethanone, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-ylethanone, 1-1-4-fluoro-phenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl-ethanone, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-1-pyrrolidinyl ethanone, usp14 inhibitor, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-yl ethanone, iu-1
CC1=CC(=C(N1C2=CC=C(C=C2)F)C)C(=O)CN3CCCC3
300.377
300.37
IU1
C18H21FN2O
JUWDSDKJBMFLHE-UHFFFAOYSA-N
1-[1-(4-fluorophenyl)-2,5-dimethylpyrrol-3-yl]-2-pyrrolidin-1-ylethanone
675434
>98%
Videos
Documents