Learn More
ITIH4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552031
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LAYSFVTPLT SMVVTKPDDQ EQSQVAEKPM EGESRNRNVH SAGAAGSRMN FRPGVLSSRQ LGLPGPPDVP DHAAYHPFRR LAILPASAPP ATSNPDPAVS RVMNMKIEET TMTTQTPA Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 36%.
ITIH4 Polyclonal antibody specifically detects ITIH4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ITIH4 | |
Polyclonal | |
Unconjugated | |
ITIH4 | |
35 kDa inter-alpha-trypsin inhibitor heavy chain H4; 70 kDa inter-alpha-trypsin inhibitor heavy chain H4; DKFZp686G21125; Gp120; H4P; IHRP; Inter alpha-trypsin inhibitor, heavy chain 4; inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein); inter-alpha-inhibitor heavy chain 4; inter-alpha-trypsin inhibitor family heavy chain-related protein; inter-alpha-trypsin inhibitor heavy chain 4; inter-alpha-trypsin inhibitor heavy chain family member 4; inter-alpha-trypsin inhibitor heavy chain family, member 4; inter-alpha-trypsin inhibitor heavy chain H4; inter-alpha-trypsin inhibitor, heavy chain-like, 1; ITI heavy chain H4; Itih4; Itih-4; ITI-HC4; ITIHL1; PK120; PK-120; Plasma kallikrein sensitive glycoprotein 120; plasma kallikrein-sensitive glycoprotein 120; PRO1851 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3700 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14624 | |
ITIH4 | |
Recombinant protein corresponding to Human ITIH4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only