missing translation for 'onlineSavingsMsg'
Learn More

ISX 9, Tocris Bioscience™

Neurogenic agent; induces neuronal differentiation of SVZ progenitors and also induces cardiomyogenic differentiation

Brand:  RD Systems (Bio-Techne) 4439/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13434764

  • £695.31 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Neurogenic agent. Mediates neuroD reporter gene induction via activation of Ca2+ influx; increases expression of neurogenic differentiation 1 (NeuroD1) transcription factor. Induces neuronal differentiation in human cortical neuronal cells (HCN), adult mouse whole brain (MWB) and subventricular zone (SVZ) progenitors. Also shown to induce cardiomyogenic differentiation of Notch-activated epicardium-derived cells in vitro.
Specifications

Chemical Identifiers

832115-62-5
234.273
isx-9, isx 9, n-cyclopropyl-5-thiophen-2-yl isoxazole-3-carboxamide, isoxazole 9 isx-9, neuronal differentiation inducer iii, n-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide, isoxazole 9, isx9, isoxazole 9 isx9, n-cyclopropyl-5-2-thienyl-3-isoxazolecarboxamide
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide
C11H10N2O2S
SYENTKHGMVKGAQ-UHFFFAOYSA-N
19582717
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3

Specifications

50mg
832115-62-5
isx-9, isx 9, n-cyclopropyl-5-thiophen-2-yl isoxazole-3-carboxamide, isoxazole 9 isx-9, neuronal differentiation inducer iii, n-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide, isoxazole 9, isx9, isoxazole 9 isx9, n-cyclopropyl-5-2-thienyl-3-isoxazolecarboxamide
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3
234.273
234.27
ISX 9
C11H10N2O2S
SYENTKHGMVKGAQ-UHFFFAOYSA-N
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide
19582717
>99%
Videos
Documents