missing translation for 'onlineSavingsMsg'
Learn More

Isradipine, Tocris Bioscience™

CaV1.x blocker

Brand:  Tocris (Bio-Techne) 2004/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13430384

  • £145.53 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective L-type voltage-gated Ca2+ channel blocker. EC50 and pA2 values are 1.4 nM and 10.3 for relaxation of depolarization- and Ca2+-induced contractions of rabbit aorta respectively; EC25 = 0.45 nM for reduction in rate of spontaneously beating guinea pig right atria. Long-acting antihypertensive agent in vivo, exerting primary effects on vascular tissue with secondary negative chronotropic action.
Specifications

Chemical Identifiers

75695-93-1
371.393
isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
3-O-methyl 5-O-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
C19H21N3O5
HMJIYCCIJYRONP-UHFFFAOYSA-N
3784
CC1=C(C(C(=C(N1)C)C(=O)OC(C)C)C2=CC=CC3=NON=C32)C(=O)OC

Specifications

10mg
75695-93-1
isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
CC1=C(C(C(=C(N1)C)C(=O)OC(C)C)C2=CC=CC3=NON=C32)C(=O)OC
371.393
371.39
Isradipine
C19H21N3O5
HMJIYCCIJYRONP-UHFFFAOYSA-N
3-O-methyl 5-O-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
3784
>99%
Videos
Documents