missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxazole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.0700g/mL |
Boiling Point | 95°C |
Flash Point | 8°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4260 to 1.428 |
Quantity | 50 g |
Beilstein | 27, 14 |
Specific Gravity | 1.07 |
Formula Weight | 69.06 |
Show More |
Safety and Handling
Product Identifier
- Isoxazole
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'