missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Spécification
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 131.52 |
Percent Purity | 90% |
Chemical Name or Material | Isoxazole-5-carbonyl chloride |
Sécurité et traitement
Identifiant produit
- Isoxazole-5-carbonyl chloride
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'