missing translation for 'onlineSavingsMsg'
Learn More

Isothiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7077730
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
288-16-4
C3H3NS
85.13
ZLTPDFXIESTBQG-UHFFFAOYSA-N
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
67515
CHEBI:35600
1,2-thiazole
C1=CSN=C1
This item is not returnable. View return policy
288-16-4
C3H3NS
85.13
ZLTPDFXIESTBQG-UHFFFAOYSA-N
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
67515
CHEBI:35600
1,2-thiazole
C1=CSN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 85.13
Percent Purity 97%
Chemical Name or Material Isothiazole
compliance-icons compliance-icons
Product Identifier
  • Isothiazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'