missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ IsoTherm-System™ IsoSafe and IsoPack Set

Used as an enzyme bar to prevent enzymes from becoming inactivated, as well as for PCR* sample preparation, sample transport and slow thawing of frozen samples.

Brand:  Eppendorf™ 3880000046

Code : 96

Additional Details : Weight : 1.08000kg

 View more versions of this product

Product Code. 10798551

  • £147.40 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

The IsoTherm™ system can be used as an enzyme bar to prevent enzymes from becoming inactivated, as well as for PCR* sample preparation, sample transport and slow thawing of frozen samples. Consists of the IsoRack™, IsoPack™ and IsoSafe™.
  • Holds 24 tubes (0.5mL/1.5mL/2.0mL Safe-Lock microcentrifuge tubes or 1.7mL Safe-Twist screw cap tubes)
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor Specialises in cooling samples effectively and constantly at -21°C or 0°C over many hours.
Specifications

Specifications

−21°C
−21°C
−21°C
IsoSafe and IsoPack Set
Videos
Documentation

Technical Documents