missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isoquinoline-5-sulfonic acid, 95%

Brand:  Alfa Aesar™ H61739.14

Code : NEW

Additional Details : CAS Number : 27655-40-9 Weight : 0.02500kg Transport : UN number : 2585 Chem class : 1a Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15468298

  • £55.50 / 25g


Estimated Shipment date
from Supplier 15-02-2018
Add to basket

Specifications

Chemical Identifiers

Isoquinoline-5-sulfonic acid
209.22
UN2585
27655-40-9
MFCD00134089

Specifications

Isoquinoline-5-sulfonic acid
25g
Solid
209.22
UN2585
>300°C
27655-40-9
95%
MFCD00134089
Videos
SDS