missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine, 99%, Thermo Scientific Chemicals

Catalog Number 10776351
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
75-31-0
C3H9N
59.11
JJWLVOIRVHMVIS-UHFFFAOYSA-N
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
6363
CHEBI:15739
propan-2-amine
CC(C)N
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
75-31-0
C3H9N
59.11
JJWLVOIRVHMVIS-UHFFFAOYSA-N
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
6363
CHEBI:15739
propan-2-amine
CC(C)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -101.0°C
Color White
Density 0.6900g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHNH2
Refractive Index 1.3725 to 1.3755
Quantity 2.5 L
Beilstein 04,152
Fieser 13,164
Merck Index 15,5255
Specific Gravity 0.69
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.3 mPa.s (20°C)
Formula Weight 59.11
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Isopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'