missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog Number 15246483
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
926-06-7
C4H10O3S
138.19
MFCD00047802
SWWHCQCMVCPLEQ-UHFFFAOYSA-N
isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b
13551
propan-2-yl methanesulfonate
CC(C)OS(=O)(=O)C
926-06-7
C4H10O3S
138.19
MFCD00047802
SWWHCQCMVCPLEQ-UHFFFAOYSA-N
isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b
13551
propan-2-yl methanesulfonate
CC(C)OS(=O)(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1450g/mL
Boiling Point 82.0°C (6.0 mmHg)
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3SO3CH(CH3)2
Refractive Index 1.4180 to 1.4210
Quantity 5 g
UN Number 1760
Beilstein 04,IV,13
Specific Gravity 1.145
Formula Weight 138.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl Methanesulfonate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropyl methanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'