missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™

Brand:  Fisher Bioreagents BP1620-10

Code : 32

Additional Details : CAS Number : 367-93-1 Weight : 0.06000kg

Quantity 10g
Packaging Glass Bottle

 View more versions of this product

Product Code. 10715114

  • £183.86 / 10g


EU Stock 9
Add to basket

Specifications

Chemical Identifiers

Isopropyl-β-D-thiogalactopyranoside
Solid
238.3
367-93-1
C9H18O5S
IPTG

Specifications

Isopropyl-β-D-thiogalactopyranoside
122°C
0.06 AU max. at 400nm, 0.13 AU max. at 300nm
Solid
238.3
Glass Bottle
−26 to −28° (+ or −)
Off-white
10g
367-93-1
C9H18O5S
15, 5126
IPTG
Single spot
Videos
SDS Documentation