missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-β-D-Thiogalactopyranoside, (Crystalline),MP Biomedicals
Click to view available options
Quantity:
1 g
5 g
Specifications
Quantity | 1 g |
IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol |
Formula Weight | 238.3 |
Physical Form | Crystalline |
Chemical Name or Material | Isopropyl-β-D-Thiogalactopyranoside |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'