missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™

Catalog Number p-151644
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
367-93-1, 123-91-1
C9H18O5S
238.30
MFCD00063273
BPHPUYQFMNQIOC-NXRLNHOXSA-N
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
656894
CHEBI:61448
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
This item is not returnable. View return policy
367-93-1, 123-91-1
C9H18O5S
238.30
MFCD00063273
BPHPUYQFMNQIOC-NXRLNHOXSA-N
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
656894
CHEBI:61448
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Color White
Loss on Drying 15% to 16% (90°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 mg
Beilstein 17, IV, 3726
Merck Index 15, 5126
Specific Rotation Condition − 27.00 (20.00°C c=1,H2O)
Specific Rotation -25° to -33° (20°C, 589nm) (c=1, water)
Formula Weight 238.3
Percent Purity ≥99.0%
Grade Biochemical
Physical Form Crystalline Powder or Needles
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'