missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
Description
- Induces the lac operon in E. coli by binding and inactivating the lac repressor.
- Selection for lac Y mutants.
- Selection for recombinant plasmids containing β-galactosidase fusion proteins.
- Induces the cellular content of lactose permease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Induces lac operon in E. coli by binding and inactivating lac repressor
- Selection for lac Y mutants
- Selection for recombinant plasmids containing beta-galactosidase fusion proteins
- Induces cellular content of lactose permea
Specifications
Melting Point | 109°C to 115°C |
Packaging | Plastic bottle |
Quantity | 1 g |
Beilstein | 4631 |
Formula Weight | 238.3 |
Percent Purity | ≥99% |
Grade | Ultrapure |
Physical Form | Powder |
Chemical Name or Material | Isopropyl-beta-D-thiogalactopyranoside (IPTG) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'