missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl chloroformate, 96%, stabilized, Thermo Scientific Chemicals

Catalog Number 10539253
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Sea freight only: For EU stock allow up to 15 days delivery.
57933-83-2
MFCD00037083
AMMGCGVWJMRTQI-UHFFFAOYSA-N
isopropenyl chloroformate, carbonochloridic acid, 1-methylethenyl ester, isopropenylchloroformate, isopropenylchloro-formate, isopropenyl carbonochloridate, isopropenyl chloridocarbonate, prop-1-en-2-yl chloroformate, chloroformic acid isopropenyl ester, chloroformic acid,isopropenyl ester, chloro prop-1-en-2-yloxy methanone
93846
prop-1-en-2-yl carbonochloridate
CC(=C)OC(=O)Cl
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Sea freight only: For EU stock allow up to 15 days delivery.
57933-83-2
MFCD00037083
AMMGCGVWJMRTQI-UHFFFAOYSA-N
isopropenyl chloroformate, carbonochloridic acid, 1-methylethenyl ester, isopropenylchloroformate, isopropenylchloro-formate, isopropenyl carbonochloridate, isopropenyl chloridocarbonate, prop-1-en-2-yl chloroformate, chloroformic acid isopropenyl ester, chloroformic acid,isopropenyl ester, chloro prop-1-en-2-yloxy methanone
93846
prop-1-en-2-yl carbonochloridate
CC(=C)OC(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1030g/mL
Boiling Point 95.0°C to 97.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5ClO2
Linear Formula CH2=C(CH3)CO2Cl
Refractive Index 1.4130 to 1.4150
Quantity 1 g
Beilstein 03,28
Fieser 08,53
Specific Gravity 1.103
Solubility Information Solubility in water: reacts with water
Stabilizer 50 to 100ppm BHT
Molecular Weight (g/mol) 120.54
Formula Weight 120.54
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Isopropenyl chloroformate, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropenyl chloroformate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'