missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Molecular Biology Grade, Fisher BioReagents

For nucleic acid precipitation

Brand:  Fisher Bioreagents BP2618-1

Code : 32

Additional Details : CAS Number : 67-63-0 Weight : 0.83500kg Transport : UN number : 1219 Chem class : 3 Pack group : II


Quantity 1L
Packaging Glass Bottle

 View more versions of this product

Product Code. 11398461

  • £22.35 / 1 L


In Stock 39
EU Stock 51
Add to basket


Description

Description

Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.
Specifications

Chemical Identifiers

Isopropanol
Molecular Biology Grade
≥99.9%
C3H8O
60.1
67-63-0
DNase-, RNase- and Protease-Free
≥99.9 %
(CH3)2CHOH
2-Propanol, IPA, Isopropyl Alcohol, sec-Propyl alcohol

Specifications

Isopropanol
67-63-0
Vapor Pressure: 43mmHg at 20°C
DNase-, RNase- and Protease-Free
≥99.9%
C3H8O
60.1
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.
DNase free
Glass Bottle
0.002% max.
1ppm max.
Pass Test
43mmHg at 20°C
2.27 mPa/s at 20°C
Colorless
7
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
0.002% max.
Molecular Biology Grade
Liquid
≥99.9 %
(CH3)2CHOH
2-Propanol, IPA, Isopropyl Alcohol, sec-Propyl alcohol
0.785g/cm³
1 ppb max.
Protease free
1.3740 to 1.3760 (at 25°C)
Pass Test
0.0001mEq/g. max.
0.05% max.
81°C
−89.5°C
1L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isopropanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documentation