missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Molecular Biology Grade, Fisher BioReagents

For nucleic acid precipitation

Brand:  Fisher Bioreagents BP2618-4

Code : 32

Additional Details : CAS Number : 67-63-0 Weight : 3.14000kg Transport : UN number : 1219 Chem class : 1 Pack group : II


Quantity 4L
Packaging Glass Bottle

 View more versions of this product

Product Code. 11308471

  • £74.47 / 4 L


In Stock 11
EU Stock 15
Add to basket


Description

Description

Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.
Specifications

Chemical Identifiers

Isopropanol
Molecular Biology Grade
≥99.9%
C3H8O
60.1
67-63-0
DNase-, RNase- and Protease-Free
≥99.9 %
(CH3)2CHOH
2-Propanol, IPA, Isopropyl Alcohol, sec-Propyl alcohol

Specifications

Isopropanol
67-63-0
Vapor Pressure: 43mmHg at 20°C
DNase-, RNase- and Protease-Free
≥99.9%
C3H8O
60.1
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.
DNase free
Glass Bottle
0.002% max.
1ppm max.
Pass Test
43mmHg at 20°C
2.27 mPa/s at 20°C
Colorless
7
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
0.002% max.
Molecular Biology Grade
Liquid
≥99.9 %
(CH3)2CHOH
2-Propanol, IPA, Isopropyl Alcohol, sec-Propyl alcohol
0.785g/cm³
1 ppb max.
Protease free
1.3740 to 1.3760 (at 25°C)
Pass Test
0.0001mEq/g. max.
0.05% max.
81°C
−89.5°C
4L
Videos
SDS Documentation