missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol, Extra Pure, SLR, Fisher Chemical™
88.56 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
88.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
26.25 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Use promo code "21741" to get your promotional price.
88.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
26.25 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Bottle
Plastic Container
Plastic-coated Glass Bottle
Description
Isopropanol, Extra Pure, SLR
Specifications
pH | 7 |
Melting Point | -89.5°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 5 L |
Packaging | Plastic Container |
Physical Form | Liquid |
Vapor Pressure | 43mmHg at 20°C |
Viscosity | 2.27 mPaS at 20°C |
Formula Weight | 60.1g/mol |
Safety and Handling
Product Identifier
- Propan-2-ol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'