missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/7490/21

40.94 GBP valid until 2021-10-30
Use promo code "14806" to get your promotional price.



Code : 21

Additional Details : CAS Number : 67-63-0 Weight : 3.93000kg Transport : UN number : 1219 Chem class : 3 Pack group : II


Packaging Plastic Bottle
Quantity 5L

 View more versions of this product

Product Code. 10070320

Quantity Price
1 £115.14 / 5 L
4 £96.49 / 5 L
16 £91.66 / 5 L
In stock, ready to ship: 198

Please to purchase this item. Need a web account? Register with us today!



For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

Isopropanol, Extra Pure, SLR

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
11674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

67-63-0
11674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1g/mol
Plastic Bottle
2.1 at 20 °C / 68 °F
81°C
-89.5°C
5L
C3H8O
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
Liquid
43mmHg at 20°C
2.27 mPaS at 20°C
Colorless
7
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isopropanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates
View accessory products
Special Offers

Special Offers

  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 14806

    2021-10-31

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/09/2021 and 31/10/2021. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions