missing translation for 'onlineSavingsMsg'
Learn More

Isoprene, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-151128
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
78-79-5, 98-29-3
C5H8
68.11
MFCD00008600
RRHGJUQNOFWUDK-UHFFFAOYSA-N
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
6557
CHEBI:35194
2-methylbuta-1,3-diene
CC(=C)C=C
78-79-5, 98-29-3
C5H8
68.11
MFCD00008600
RRHGJUQNOFWUDK-UHFFFAOYSA-N
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
6557
CHEBI:35194
2-methylbuta-1,3-diene
CC(=C)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -146.0°C
Color Colorless to Yellow
Density 0.6800g/mL
Boiling Point 34.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(CH3)=CH2
Refractive Index 1.4205 to 1.4225
Quantity 100 mL
Beilstein 01, 252
Fieser 13,196
Merck Index 15, 5247
Specific Gravity 0.68
Solubility Information Solubility in water: 0.7g/L (25°C). Other solubilities: miscible with alcohol,ether and acetone,oils/fat
Stabilizer 100 to 250ppm 4-tert-butylcatechol
Viscosity 0.225 mPa.s (15°C)
Formula Weight 68.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isoprene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'