Learn More
Merck Isopore™ Membrane Filters, 8 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
£132.00 - £282.00
Specifications
Airflow Rate | 60 L/min/ cm2/psi |
---|---|
Color | White |
Filter Type | Screen Filter |
Material | Polycarbonate |
Pore Size | 8 μm |
Product Code | Brand | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
10703131
|
Merck
TETP01300 |
13 mm |
£132.00
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10044510
|
Merck
TETP02500 |
25 mm |
£195.00
Pack of 100 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
10459840
|
Merck
TETP04700 |
47 mm |
£282.00
Pack of 100 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
60 L/min/ cm2/psi | |
Screen Filter | |
8 μm | |
For analysis of airborne contaminants & other particles using optical or electron microscopy | |
100Pack | |
140°C | |
Filter Membranes |
White | |
Polycarbonate | |
TETP | |
5 to 20% | |
Plain | |
18 μm |