Learn More
Merck Isopore™ Membrane Filters, 0.2 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
£125.00 - £535.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Filter Type | Screen Filter |
Material | Polycarbonate |
Pore Size | 0.22 μm |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) | Color | Flow Rate | Bubble Point | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Color | Flow Rate | Bubble Point | Price | Quantity & Availability | ||
10084440
|
Merck
GTTP01300 |
13 mm | White | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi | 50 psi |
£143.00
Pack of 100 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||
10702731
|
Merck
GTBP01300 |
13 mm | Black | Avg. 16 mL/min x cm2 (typical results @ 10 psi);6 mL/min x cm2/psi | 50 psi |
£125.00
Pack of 100 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||
10712381
|
Merck
GTTP02500 |
25 mm | White | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi | 50 psi |
£220.00
Pack of 100 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
||
10527092
|
Merck
GTBP02500 |
25 mm | Black | Avg. 16 mL/min x cm2 (typical results @ 10 psi);6 mL/min x cm2/psi | 50 psi |
£287.00
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
10722381
|
Merck
GTTP03700 |
37 mm | White | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi | 50 psi |
£214.00
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
10696721
|
Merck
GTTP04700 |
47 mm | White | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi | 50 psi |
£282.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10742381
|
Merck
GTBP04700 |
47 mm | Black | Avg. 16 mL/min x cm2 (typical results @ 10 psi);6 mL/min x cm2/psi | 50 psi |
£535.00
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
10606731
|
Merck
GTTP14250 |
142 mm | White | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi | 50 psi |
£499.00
Pack of 50 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
10300462
|
Merck
GTTP09030 |
90 mm | White | 0.007 L/min/ cm2/psi (Air), 16 mL/min/ cm2 (water) | ≥5.2 bar |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
Polycarbonate | |
Non-sterile | |
Hydrophilic |
Screen Filter | |
0.22 μm | |
Filter Membranes |