missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isopentyl dodecanoate, 97%

Brand:  Alfa Aesar™ 042022.AP

Code : NEW

Additional Details : CAS Number : 6309-51-9 Weight : 0.01000kg

Quantity 500mL

 View more versions of this product

Product Code. 11365959

  • £180.31 / 500mL


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Isopentyl dodecanoate
270.46
Dodecanoic acid isopentyl ester; Isoamyl laurate
6309-51-9
MFCD00048429

Specifications

Isopentyl dodecanoate
6309-51-9
Liquid
270.46
Dodecanoic acid isopentyl ester; Isoamyl laurate
164° (327°F)
500mL
0.86g/mL
97%
MFCD00048429
1.436
Videos
SDS