missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopentyl Acetate, Reagent grade, 98%, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -78°C |
Color | Colorless |
Density | 0.876g/mL at 25°C |
Boiling Point | 142°C |
Explosion Limit | 0.075 |
Flash Point | 25°C (77°F) |
Ignition Point | 680°F |
Packaging | Stainless Steel Can |
Linear Formula | CH3COOCH2CH2CH(CH3)2 |
Refractive Index | n20/D 1.4(lit.) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'