missing translation for 'onlineSavingsMsg'
Learn More

Isonipecotic Acid Hydrochloride 97%, ACROS Organics™

Brand:  Acros Organics 124080250

Code : 29

Additional Details : CAS Number : 5984-56-5 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10316480

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Isonipecotic acid hydrochloride
5984-56-5
97%
165.62
4-Piperidinecarboxylic acid, hydrochloride
97%
≥96.0%
C6H11NO2·HCl
MFCD00044736

Specifications

Isonipecotic acid hydrochloride
97%
Crystalline Powder
97%
165.62
14, 5189
4-Piperidinecarboxylic acid, hydrochloride
0.5% max. (K.F.)
25g
5984-56-5
≥96.0%
C6H11NO2·HCl
MFCD00044736
Authentic
Glass bottle
White to cream
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isonipecotic acid hydrochloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS