missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinic acid, 99%, Thermo Scientific Chemicals
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 310°C to 315°C |
Color | Cream |
Boiling Point | 260°C (15.0 mmHg) |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 100 g |
Beilstein | 22,45 |
Merck Index | 15,5233 |
Solubility Information | Solubility in water: 5.2g/L (20°C). Other solubilities: practically insoluble in benzene,ether and,boiling alcohol |
Formula Weight | 123.11 |
Show More |
Safety and Handling
Product Identifier
- Isonicotinic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'