missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 310°C to 315°C |
Color | Cream |
Boiling Point | 260°C (15.0 mmHg) |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 2.5 kg |
Beilstein | 22,45 |
Merck Index | 15,5233 |
Solubility Information | Solubility in water: 5.2g/L (20°C). Other solubilities: practically insoluble in benzene,ether and,boiling alcohol |
Formula Weight | 123.11 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'