missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline, 97%, Thermo Scientific Chemicals

Catalog Number 10491294
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
496-12-8
C8H9N
119.17
MFCD00605324
GWVMLCQWXVFZCN-UHFFFAOYSA-N
isoindoline, 1h-isoindole, 2,3-dihydro, iso-indoline, 2-azaindan, 1,2-dihydroisoindole, 1,3-dihydroisoindole, dihydroisoindole, pubchem16248, acmc-1am9e
422478
2,3-dihydro-1H-isoindole
C1C2=CC=CC=C2CN1
This item is not returnable. View return policy
496-12-8
C8H9N
119.17
MFCD00605324
GWVMLCQWXVFZCN-UHFFFAOYSA-N
isoindoline, 1h-isoindole, 2,3-dihydro, iso-indoline, 2-azaindan, 1,2-dihydroisoindole, 1,3-dihydroisoindole, dihydroisoindole, pubchem16248, acmc-1am9e
422478
2,3-dihydro-1H-isoindole
C1C2=CC=CC=C2CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17°C
Color White
Density 1.0500g/mL
Boiling Point 221°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5700 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.05
Formula Weight 119.17
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Isoindoline
Show More Show Less
compliance-icons
Product Identifier
  • Isoindoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'