Isohexane, contains <5% n-Hexane, for HPLC, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 383810010

1LT Isohexane, for HPLC, contains <5% n-Hexane

UNSPSC: 12190000

Code: 50

Additional Details:
Additional Details: CAS Number: 107-83-5 Weight: 0.71000kg Transport: UN number : 1208 Chem class : 1 Pack group : II


Quantity 1L

Product Code. 10776373

Quantity Price
1 £ 162.7 / 1 L
Estimated Shipment
03-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Isohexane
CAS 73513-42-5
Molecular Formula C6H14
Formula Weight 86.18
MDL Number MFCD00009406

Specification

Formula Weight 86.18
Quantity 1L
Boiling Point 60.0°C
Melting Point −154.0°C
Flash Point −23°C
Density 0.66g/cm3
CAS 73513-42-5
MDL Number MFCD00009406
Molecular Formula C6H14
Chemical Name or Material Isohexane
Documentation