Isohexane, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 383820010

1LT Isohexane, for pesticide residue analysis, contains <5% n-Hexane

UNSPSC: 12352100

Code: 50

Additional Details:
Additional Details: CAS Number: 107-83-5 Weight: 0.65300kg Transport: UN number : 1208 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 1L

Product Code. 10598463

Quantity Price
1 £ 36.1 / 1 L
Estimated Shipment
27-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Isohexane
Name Note for residue and pesticides analysis, >5% n-Hexane
CAS 73513-42-5
Purity Grade For residue analysis
Molecular Formula C6H14
Linear Formula CH3CH2CH2CH(CH3)2
Formula Weight 86.18

Specification

Physical Form Liquid
Formula Weight 86.18
Quantity 1L
Color Clear
Linear Formula CH3CH2CH2CH(CH3)2
Nitrogen and Phosphorus Impurities Passes Test
Residue after Evaporation 2ppm max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Purity Grade For residue analysis
Flame Ionisable Impurities Passes Test
CAS 73513-42-5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Water 0.01% max. (coulometric)
Density 0.6600g/mL
Name Note for residue and pesticides analysis, >5% n-Hexane
Molecular Formula C6H14
Health Hazard 1 Danger
Chemical Name or Material Isohexane
Electron Capturing Impurities Passes Test
Documentation Safety and Handling

Danger

GHS H Statement:
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.