missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoguvacine hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Isoguvacine·HCl is an agonist of γ-amino butyric acid receptor. Isoguvacine interactions have many similar biochemical features of GABA synaptic function. Isoguvacine·HCl is a specific GABAA receptor agonist. This compound supp
Isoguvacine·HCl is an agonist of γ-amino butyric acid receptor. Isoguvacine interactions have many similar biochemical features of GABA synaptic function. Isoguvacine·HCl is a specific GABAA receptor agonist. This compound supp
Specifications
CAS | 64603-90-3 |
Color | White |
Molecular Formula | C6H10ClNO2 |
MDL Number | MFCD01365699 |
Quantity | 100 mg |
Synonym | isoguvacine, 1,2,3,6-tetrahydro-pyridine-4-carboxylic acid, 1,2,3,6-tetrahydro-4-pyridinecarboxylic acid, unii-ytf580771y, chembl39071, 4-pyridinecarboxylic acid, 1,2,3,6-tetrahydro, 1,2,3,6-tetrahydropyridinium-4-carboxylate, 1,2,3,6-tetrahydropyridin-1-ium-4-carboxylate, artemisinin sp, spectrum_001974 |
Solubility Information | Soluble in water |
InChI Key | SUWREQRNTXCCBL-UHFFFAOYSA-N |
SMILES | [H+].[Cl-].OC(=O)C1=CCNCC1 |
IUPAC Name | 1,2,3,6-tetrahydropyridine-4-carboxylic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'