missing translation for 'onlineSavingsMsg'
Learn More

Isochroman, 99%, Thermo Scientific Chemicals

Catalog Number 10498270
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
493-05-0
C9H10O
134.18
MFCD00006913
HEBMCVBCEDMUOF-UHFFFAOYSA-N
isochroman, isochromane, 3,4-dihydro-1h-2-benzopyran, 1h-2-benzopyran, 3,4-dihydro, 3,4-dihydro-1h-benzo c pyran, pubchem20553, acmc-209kfu, isochroman 10g, ksc237m1l
96266
CHEBI:33225
3,4-dihydro-1H-isochromene
C1CC2=CC=CC=C2CO1
This item is not returnable. View return policy
493-05-0
C9H10O
134.18
MFCD00006913
HEBMCVBCEDMUOF-UHFFFAOYSA-N
isochroman, isochromane, 3,4-dihydro-1h-2-benzopyran, 1h-2-benzopyran, 3,4-dihydro, 3,4-dihydro-1h-benzo c pyran, pubchem20553, acmc-209kfu, isochroman 10g, ksc237m1l
96266
CHEBI:33225
3,4-dihydro-1H-isochromene
C1CC2=CC=CC=C2CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 75°C (4.0 Mbar)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.546
Quantity 25 g
Beilstein 17, 53
Specific Gravity 1.06
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isochroman, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'