missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7027598
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
79-30-1
C4H7ClO
106.549
MFCD00000717
DGMOBVGABMBZSB-UHFFFAOYSA-N
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
62325
2-methylpropanoyl chloride
CC(C)C(=O)Cl
This item is not returnable. View return policy
79-30-1
C4H7ClO
106.549
MFCD00000717
DGMOBVGABMBZSB-UHFFFAOYSA-N
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
62325
2-methylpropanoyl chloride
CC(C)C(=O)Cl

Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

Solubility
Miscible with chloroform, glacial acetic acid, ether, toluene, methylene chloride and benzene.

Notes
Moisture sensitive. Incompatible with bases, water, strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY
Melting Point ∽-90°C
Density 1.017
Boiling Point 91°C to 93°C
Flash Point 8°C (46°F)
Assay Percent Range 0.98
Refractive Index 1.407
Quantity 100 g
UN Number UN2395
Beilstein 605393
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform,glacial acetic acid,ether,toluene,methylene chloride and benzene.
Formula Weight 106.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'