Isobutyltrichlorosilane, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 338060050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 18169-57-8 Weight: 0.05575kg Transport: UN number : 2986 Chem class : 1a Pack group : II


Packaging Glass Bottle
Quantity 5mL

Product Code. 10663183

Quantity Price
1 £ 11.0 / 5mL
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Isobutyltrichlorosilane
Name Note 98%
CAS 18169-57-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H9Cl3Si
Linear Formula (Cl3)SiCHCH(CH3)
Formula Weight 191.56
MDL Number MFCD00053185

Specification

Formula Weight 191.56
Physical Form Liquid
Assay ≥97.5%
Quantity 5mL
Color Clear colorless to slightly yellow
EINECS Number 242-053-4
Linear Formula (Cl3)SiCHCH(CH3)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 18169-57-8
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00053185
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
Density 1.15g/mL
Name Note 98%
Molecular Formula C4H9Cl3Si
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Isobutyltrichlorosilane
TSCA TSCA
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.