missing translation for 'onlineSavingsMsg'
Learn More

Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4089710
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
5674-02-2, 60-29-7
116.87
MFCD00000471
CHEBI:75272
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
5674-02-2, 60-29-7
116.87
MFCD00000471
CHEBI:75272

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8300g/mL
Flash Point −040°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H9ClMg
Linear Formula (CH3)2CHCH2MgCl
Quantity 100 mL
Specific Gravity 0.83
Synonym splitomicin, 1,2-dihydro-3h-naphtho 2,1-b pyran-3-one, 1,2-dihydro-3h-benzo f chromen-3-one, 1h-benzo f chromen-3 2h-one, chembl86537, 1,2-dihydrobenzo f chromen-3-one, splitomycin, tocris-1542, d09suo, splitomicin hplc , powder
Solubility Information Solubility in water: reacts.
InChI Key ISFPDBUKMJDAJH-UHFFFAOYSA-N
SMILES C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
IUPAC Name 1,2-dihydrobenzo[f]chromen-3-one
PubChem CID 5269
Formula Weight 116.87
Chemical Name or Material Isobutylmagnesium chloride, 2M solution in diethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutylmagnesium chloride, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'