missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutylhydrazine hydrochloride, 97%

Brand:  Alfa Aesar™ H64287.MD

Code : NEW

Additional Details : CAS Number : 237064-47-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15491409

  • £17.38 / 250mg


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Applications
It finds its application in the discovery and characterization of mapk-activated protein kinase-2 prevention of activation inhibitors.

Solubility
Slightly soluble in water. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

Isobutylhydrazine hydrochloride
(CH3)2CHCH2NHNH2·HCl
MFCD06245510
237064-47-0
124.61
2-Methylpropylhydrazine hydrochloride

Specifications

Isobutylhydrazine hydrochloride
237064-47-0
97%
124.61
2-Methylpropylhydrazine hydrochloride
250mg
Solid
(CH3)2CHCH2NHNH2·HCl
MFCD06245510
Videos
SDS