missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene oxide, 99+%, Thermo Scientific Chemicals

Catalog Number p-7045196
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
558-30-5
C4H8O
72.107
MFCD00066354
GELKGHVAFRCJNA-UHFFFAOYSA-N
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
11208
2,2-dimethyloxirane
CC1(CO1)C
This item is not returnable. View return policy
558-30-5
C4H8O
72.107
MFCD00066354
GELKGHVAFRCJNA-UHFFFAOYSA-N
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
11208
2,2-dimethyloxirane
CC1(CO1)C

Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.

Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.808
Boiling Point 51°C to 52°C
Flash Point −32°C (−25°F)
Odor Ether-like
Refractive Index 1.375
Quantity 5 g
UN Number UN2924
Beilstein 102408
Solubility Information Miscible with ethanol and diethyl ether. Slightly miscible with water.
Formula Weight 72.11
Percent Purity ≥99%
Chemical Name or Material Isobutylene oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'