missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number p-7051777
Click to view available options
Quantity:
250 mL
1 L
4 L
This item is not returnable. View return policy
97-86-9
C8H14O2
142.20
MFCD00008931,MFCD00084435
RUMACXVDVNRZJZ-UHFFFAOYSA-N
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
7352
2-methylpropyl 2-methylprop-2-enoate
CC(C)COC(=O)C(C)=C
This item is not returnable. View return policy
97-86-9
C8H14O2
142.20
MFCD00008931,MFCD00084435
RUMACXVDVNRZJZ-UHFFFAOYSA-N
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
7352
2-methylpropyl 2-methylprop-2-enoate
CC(C)COC(=O)C(C)=C

Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.889 g/mL
Boiling Point 155°C
Flash Point 46°C (115°F)
Quantity 250 mL
UN Number UN2283
Beilstein 1747595
Solubility Information Soluble in water at 20°C,2g/L
Formula Weight 142.2
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Isobutyl methacrylate, Stabilized with 10ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'