missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutyl dodecanoate, 99+%

Brand:  Alfa Aesar™ 042023.AC

Code : NEW

Additional Details : CAS Number : 37811-72-6 Weight : 0.01000kg

Quantity 25mL

 View more versions of this product

Product Code. 11335047

  • £19.29 / 25mL


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Isobutyl dodecanoate
256.43
Isobutyl laurate
37811-72-6
MFCD00048424

Specifications

Isobutyl dodecanoate
37811-72-6
Liquid
256.43
Isobutyl laurate
25mL
0.86g/mL
≥99%
MFCD00048424
150° (302°F)
Videos
SDS