missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog Number p-7022084
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
543-27-1
C5H9ClO2
136.575
MFCD00000642
YOETUEMZNOLGDB-UHFFFAOYSA-N
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
62365
2-methylpropyl carbonochloridate
CC(C)COC(=O)Cl
This item is not returnable. View return policy
543-27-1
C5H9ClO2
136.575
MFCD00000642
YOETUEMZNOLGDB-UHFFFAOYSA-N
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
62365
2-methylpropyl carbonochloridate
CC(C)COC(=O)Cl

Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.
TRUSTED_SUSTAINABILITY
Melting Point °C to 80°C
Density 1.048
Boiling Point 128°C to 129°C
Flash Point 30°C (86°F)
Assay Percent Range 98%
Linear Formula CO2CH2CH(CH3)2
Refractive Index 1.406
Quantity 25 g
UN Number UN2742
Beilstein 956590
Sensitivity Moisture sensitive
Merck Index 14,5139
Solubility Information Immiscible with water.
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl Chloroformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'