missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutanesulfonyl chloride, 97%

Brand:  Alfa Aesar™ H64378.03

Code : NEW

Additional Details : CAS Number : 35432-36-1 Weight : 0.00100kg Transport : UN number : 3265 Chem class : 1a Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15441559

  • £48.60 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

Isobutanesulfonyl chloride
(CH3)2CHCH2SO2Cl
MFCD07369800
1748728
35432-36-1
156.62
UN3265
2-Methyl-1-propylsulfonyl chloride

Specifications

Isobutanesulfonyl chloride
1g
1.20g/mL
97%
156.62
UN3265
2-Methyl-1-propylsulfonyl chloride
Colorless
Moisture Sensitive
35432-36-1
Liquid
(CH3)2CHCH2SO2Cl
MFCD07369800
1748728
1.4540
Videos
SDS