missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Iso-Versinic™ Reference Tubing for MF1 and MF4 Pump Heads

For use with high chemical resistance applications

Brand:  Gilson™ F1817745

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10643305

  • £102.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • so-Versinic™
Specifications

Specifications

Yes
Black
Iso-Versinic
40cm
For MF1 and MF4 Pump Heads
–20°C
Acids
2mm
24mL/min.
Pump Tubing
Where high chemical resistance is required
Impermeable to most gases
4Pack
Videos