missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISLR-2 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | ISLR-2 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590 Accession # Q6UXK2 |
Gene ID (Entrez) | 57611 |
Gene Accession No. | Q6UXK2 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
ISLR-2 Polyclonal specifically detects ISLR-2 in Human samples. It is validated for Western Blot.Specifications
ISLR-2 | |
57611 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human ISLR-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human ISLR-2 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590 Accession # Q6UXK2 | |
Q6UXK2 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Sheep | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only