missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Isamoltane hemifumarate
5-HT1B antagonist
POA
Specifications
Assay Percent Range | >99% |
---|---|
Chemical Name or Material | 1-[(1-Methylethyl)amino]-3-[2-(1H-pyrrol-1-yl)]-propan-2-ol hemifumarate |
CAS | 874882-92-5 |
Description
5-HT1B antagonist, approximately 30-fold selective over 5-HT1A. Also possesses affinity for β-adrenergic receptors.Specifications
>99% | |
1-[(1-Methylethyl)amino]-3-[2-(1H-pyrrol-1-yl)]-propan-2-ol hemifumarate | |
874882-92-5 |
For Research Use Only