Learn More
Tocris Bioscience™ Isamoltane hemifumarate
Brand:  Tocris Bioscience™ 099210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13488243
  • £128.25 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
5-HT1B antagonist, approximately 30-fold selective over 5-HT1A. Also possesses affinity for β-adrenergic receptors.
Specifications
Specifications
1-[(1-Methylethyl)amino]-3-[2-(1H-pyrrol-1-yl)]-propan-2-ol hemifumarate
>99%
Observed MW: 332.4
Soluble to 10mM with gentle warming in water
874882-92-5
C16H22N2O2.½C4H4O4