Learn More
IRX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563000
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SAEDEGISLH VDSLTDHSCS AESDGEKLPC RAGDPLCESG SECKDKYDDL EGDED Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
IRX2 Polyclonal antibody specifically detects IRX2 in Human samples. It is validated for Immunohistochemistry.Specifications
IRX2 | |
Polyclonal | |
Unconjugated | |
IRX2 | |
homeodomain protein IRXA2; iroquois homeobox 2; iroquois homeobox protein 2; Iroquois related homeobox 2 (Drosophila); iroquois-class homeobox protein Irx6; Iroquois-class homeodomain protein IRX-2; Irx2; Irx6; IRXA2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZI1 | |
IRX2 | |
Recombinant protein corresponding to Human IRX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only