missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, >99% (metals basis), Alfa Aesar™

Form: approx. 25g/m

Brand:  Alfa Aesar 014246.J2

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.01000kg

Quantity 5 x 50m

 View more versions of this product

Product Code. 11387817

  • £294.00 / Pack of 5


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

Iron
Wire, 2mm (0.08 in.) dia.
Fe
7439-89-6
>99% (Metals basis)
MFCD00010999

Specifications

Iron
7439-89-6
Wire, 2mm (0.08 in.) dia.
Fe
Gray
5 x 50m
Gray wire
>99% (Metals basis)
MFCD00010999
Videos
SDS