missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron wire, 1.0mm (0.04in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
61.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 cm
250 cm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Assay Percent Range | 99.99% (metals basis) |
Quantity | 50 cm |
Chemical Name or Material | Iron wire |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'