missing translation for 'onlineSavingsMsg'
Learn More

Iron sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407466
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12068-85-8
FeS2
119.97
MFCD00064690
NFMAZVUSKIJEIH-UHFFFAOYSA-N
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
14788
S=[Fe]=S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12068-85-8
FeS2
119.97
MFCD00064690
NFMAZVUSKIJEIH-UHFFFAOYSA-N
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
14788
S=[Fe]=S

Iron sulfide is used in photovoltaics and lithium ion battery cathodes. It is also studied as a potential material for the sequestration of environmental contaminants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1171°C
Color Gray
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name bis(sulfanylidene)iron
Formula Weight 119.98
Odor Odorless
Appearance Gray powder
Density 4.87
Chemical Name or Material Iron sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'